Scalpel Handles, Integra
Supplier: Miltex
4-10 4-11 4-12 4-16 4-17 4-18 4-20 4-400 4-401 4-405 4-7R 4-9
76470-852EA 129.13 CAD
76470-852 76470-854 76470-856 76470-858 76470-860 76470-862 76470-864 76470-866 76470-868 76470-870 76470-872 76470-874
Scalpel Handles, Integra
Scalpels and Scalpel Assemblies Scalpel Handles
EZ-OFF reusable plastic knife handles.
  • Autoclavable
  • Quick and easy removal of the blade
Fits mini blade (blades not available from Integra Miltex).