Q5U Hot Start High-Fidelity DNA Polymerase, New England Biolabs
Supplier: New England Biolabs (NEB)
M0515L
M0515S
CA76339-738EA
1119
CAD
CA76339-738
CA76339-740
Q5U Hot Start High-Fidelity DNA Polymerase, New England Biolabs
Nucleic Acid Reagents
End-point PCR Enzymes and Kits
Q5U Hot Start high-fidelity DNA Polymerase is a modified version of Q5 high-fidelity DNA Polymerase, a novel thermostable DNA polymerase that possesses 3' to 5' exonuclease activity, and is fused to a processivity-enhancing Sso7d domain.
- Achieve superior amplification of bisulfite-converted, deaminated, or damaged DNA (FFPE)
- Prevent carryover contamination when combined with dUTP and Uracil DNA Glycosylase (UDG) treatment
- Generate higher assembly efficiency and improve accuracy in USER cloning
- Enable room temperature setup with aptamer-based hot start formulation
Q5U contains a mutation in the uracil-binding pocket that enables the ability to read and amplify templates containing uracil and inosine bases.