Signs, 'WARNING, DO NOT UNPLUG THE REFRIGERATOR/FREEZER OR BREAK CIRCUIT, EXPENSIVE VACCINE IN STORAGE', Accuform®
Supplier: ACCUFORM MANUFACTURING, INC
MBDX300VS MBDX300VP MBDX300VA
76471-236EA 15.18 CAD
76471-236 76471-238 76471-240 76471-242 76471-244 76471-246
Signs, 'WARNING, DO NOT UNPLUG THE REFRIGERATOR/FREEZER OR BREAK CIRCUIT, EXPENSIVE VACCINE IN STORAGE', Accuform®
Signs
Vaccination signs communicate health and safety policies, reinforce procedures, and identify locations.
  • Multiple material and size options available
Choose from three material options. Adhesive Vinyl signs feature a printed matte surface with permanent acrylic adhesive back. These are moisture-resistant for a variety of light-duty indoor and sheltered outdoor marking needs. Plastic signs are durable with slight flex. Plastic is a good material choice for indoor and sheltered outdoor environments, as the printed surface has clear UV protective overcoat. Aluminum signs provide the strength of metal without rusting. They feature a printed surface protected by UV coating to withstand splashes, wash-downs, and outdoor conditions. They also offer abrasion and chemical resistance.