Hi-T4 DNA Ligase
Supplier: New England Biolabs (NEB)
M2622L
M2622S
CA76407-352EA
442
CAD
CA76407-352
CA76407-354
Hi-T4 DNA Ligase
Enzymes
A thermotolerant variant of T4 DNA Ligase, Hi-T4 DNA Ligase catalyzes the formation of a phosphodiester bond between juxtaposed 5 phosphate and 3 hydroxyl termini in duplex DNA or RNA and is designed to function at higher temperatures than wild type T4 DNA Ligase.
- Hi-T4 DNA Ligase is an engineered variant of T4 DNA Ligase with improved thermostability
- Capable of joining blunt end and cohesive end termini
- Capable of repairing single stranded nicks in duplex DNA and some DNA/RNA hybrids
- Supplied with both T4 DNA Ligase buffer and StickTogether DNA Ligase buffer
Hi-T4 DNA Ligase will join blunt end and cohesive end termini as well as repair single-stranded nicks in duplex DNA, RNA or DNA/RNA hybrids at temperatures as high as 50 °C.