OneTaq Quick-Load DNA Polymerase, New England Biolabs
Supplier: New England Biolabs (NEB)
M0509L M0509X
CA76284-674EA 154 CAD
CA76284-674 CA76284-676 CA76284-678
OneTaq Quick-Load DNA Polymerase, New England Biolabs
Nucleic Acid Reagents End-point PCR Enzymes and Kits
OneTaq Quick-Load DNA Polymerase is an optimized, ready-to-use blend of Taq and Deep Vent DNA Polymerases ideally suited to routine PCR applications from a variety of templates, including pure DNA solutions, bacterial colonies and cDNA products.
  • Includes green Quick-Load reaction buffer for easy and direct loading of PCR products onto gels
  • 2X higher fidelity than TaqCompatible with standard Taq protocols
The 3' to 5' exonuclease activity of Deep Vent DNA Polymerase increases the fidelity and robust amplification of Taq DNA Polymerase. The convenient quick-load buffer contains two commonly used tracking dyes for direct loading of PCR products on DNA gels. On a 1% agarose gel in 1X TBE, Xylene Cyanol FF migrates at ~4 kb and Tartrazine migrates at ~10 bp. Both dyes are present in concentrations that do not mask comigrating DNA bands.