SensiCare® Neoprene Surgical Gloves, Medline
Supplier: Medline
MSG6355 MSG6360 MSG6365 MSG6370 MSG6375 MSG6380 MSG6385 MSG6390
76472-720BX 263.4 CAD
76472-720 76472-722 76472-724 76472-726 76472-728 76472-730 76472-732 76472-734
SensiCare® Neoprene Surgical Gloves, Medline
Gloves
SensiCare® Neoprene gloves offer a safe and economical alternative to natural rubber latex. Crafted from a second generation neoprene that exhibits outstanding performance characteristics.
  • Smooth finish designed for tactile feel
  • Brown color helps reduce glare from overhead lights in operating room
  • Designed to enhance comfort
  • Chemotherapy tested ASTM D6978-05
SensiCare® Neoprene surgical gloves provide industry-leading chemotherapy protection and a safe, economical alternative to natural rubber latex. Manufactured with an improved polycholorpreneMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
Latex/Latex-free Latex-free
Powdered/Powder-free Powder-free
Finish Smooth
Cuff style Beaded
Finger thickness 7.8 mil
Sterility Sterile