SP Bel-Art Stainless Steel Economy Scoops, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F36739-1000 F36739-2000
76459-124EA 72.13 CAD
76459-124 76459-126 76459-128
SP Bel-Art Stainless Steel Economy Scoops, Bel-Art Products, a part of SP
Scoops
Low-cost, sturdy scoop ideal for general laboratory use with high sides to prevent spillage.
  • Reusable, high-quality scoops help reduce laboratory waste
  • Useful in applications with limited salt exposure
  • 304 grade stainless steel
  • Autoclavable
Helps to meet sustainability goals when used in place of disposable scoops.