StaticCare™ ESD Cut Resistant Gloves, Transforming Technologies
Supplier: Transforming Technologies
GL2501 GL2502 GL2503 GL2504 GL2505 GL2506 GL2501T GL2502T GL2503T GL2504T GL2505T GL2506T GL2501P GL2502P GL2503P GL2504P GL2505P GL2506P
76472-900PK 319.74 CAD
76472-900 76472-902 76472-904 76472-906 76472-908 76472-910 76472-912 76472-914 76472-916 76472-918 76472-920 76472-922 76472-924 76472-926 76472-928 76472-930 76472-932 76472-934
StaticCare™ ESD Cut Resistant Gloves, Transforming Technologies
Gloves
The StaticCare™ ESD Cut Resistant Gloves combine powerful cut resistance, comfort, and uniform ESD protection into one superior performing glove. The gloves are certified ANSI Cut Level 3 while maintaining softness, flexibility, and comfort.
  • Multi-purpose electronics inspection and handling glove
  • Typical ESD properties: <10⁷ ohms
  • UHMWPE construction with conductive carbon
  • Available uncoated, fingertip coated, or palm coated
  • Finished knit wrist cuff
The StaticCare™ GL2500 series gloves provide superior ESD protection because conductive carbon threads are combined with the cut resistant fibers to provide uniform static decay rates.The gloves are constructed with 18 gauge Ultra High Molecular Weight Polyethylene (UHMWPE) thread which provides great cut protection, as well as outstanding abrasion and tear resistance. They are rated ANSI Cut Level 3, which offers a basic level protection against cuts or punctures.
A finished knitwrist cuff provides a snug fit to reduce slippage and keep debris out. Available with polyurethane coated finger tips or palms to improve grip while maintaining breathability. The gloves may be laundered for repeated use.
Certifications: ASTM ANSI Cut Level 3.
Packaging: Pack of 12 pairs.