VWR® Sterile Standard Square Weighing Boats, White
Supplier: VWR International
76472-584 76540-490
76472-584CS 990.4 CAD
76472-584 76540-490
VWR® Sterile Standard Square Weighing Boats, White
Boats Weighing Boats
These square weighing boats are flexible and feature smooth surfaces to provide accurate pour-outs with minimal sample loss.
  • Electron beam sterilized to reduce the risk of cross-contamination and compromised samples
  • Made from 100% virgin anti-static polystyrene that is waterproof and reduces the chance of materials sticking to the units
  • Disposable
  • Can be used with microwave moisture analyzers
  • Rounded corners simplify transfer and allow flexing for controlled pouring of liquid, powdered, or granular samples
  • Boats are shallow with a wide, flat bottom to resist tipping
  • FDA/food-grade compliant
Ideal for applications where there is concern of cross-contamination and ‘clean’ is a necessity in life science research, food, bio-pharmaceutical, environmental & molecular research environments.
Caution: Expires 3 years from the date of sterilization.