VWR® Eco Racks
Supplier: VWR International
76051-610 76051-612
76051-610EA 131.45 CAD
76051-610 76051-612
VWR® Eco Racks
Racks Cryogenic Box Racks
Eco racks for 3" boxes.
  • Stainless steel
  • Double welds
  • Smoothed edges
  • Lifetime warranty
Freezer Racks are made to the highest levels of quality with pharmaceutical grade stainless steel, double welds, smoothed edges and an unconditional lifetime warranty.