Multi-Channel Pipettes, Adjustable Spacing, Move It®, Eppendorf
Supplier: Eppendorf
3125000150 3125000168 3125000176 3125000184 3125000192 3125000206 3125000150 3125000168 3125000176 3125000184 3125000192 3125000206 3125000117 3125000125 3125000133 3125000141
76452-826EA 3930.52 CAD
76452-826 76452-828 76452-830 76452-832 76452-834 76452-836 CA76474-580 CA76474-582 CA76474-584 CA76474-586 CA76474-588 CA76474-590 76627-818 76627-816 76627-814 76627-820
Multi-Channel Pipettes, Adjustable Spacing, Move It®, Eppendorf
Pipettes Micropipettes
Move It® is an adjustable spacing multichannel pipette that allows easy format changes with 70% time-savings compared to single-channel pipettes.
  • For plates (384, 96, 48, 24 and 12-well), tubes (1.5 and 2.0 ml), and agarose gels
  • Tip spacing freely selectable from 4.5 to 33 mm, depending on pipette variant
  • Spacing control for quick switches backwards and forwards between source and destination formats
  • Manual adjustment knob for smooth vibration-free format switches and liquid transfers without dripping
  • Tubeless system for reliability and safety
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact
Environmental certification Renewable Energy Certificate, ISO 14001 Renewable Energy Certificate, ISO 14001 Renewable Energy Certificate, ISO 14001