NEBExpress Cell-Free E. coli Protein Synthesis System, New England Biolabs
Supplier: New England Biolabs (NEB)
E5360L
E5360S
CA76407-362EA
2891
CAD
CA76407-362
CA76407-364
NEBExpress Cell-Free E. coli Protein Synthesis System, New England Biolabs
Nucleic Acid Reagents
Nucleic Acid Purification Kits and Reagents
The NEBExpress Cell-free E. coli Protein Synthesis System is an extract-based transcription/translation system derived from E. coli cells that have been engineered for high in vitro synthesis performance. It is designed to synthesize proteins ranging in size from 17 to 230 kDa in just 2 to 4 hours under the control of T7 RNA Polymerase.
- Synthesize high yields of protein (typically 0.5 mg/ml)
- Protein can be synthesized and visualized in approximately 2 to 4 hours
- Templates can be plasmid DNA, linear DNA or mRNA