BclI-HF, New England Biolabs
Supplier: New England Biolabs (NEB)
R3160L R3160S
103501-474EA 464.52 CAD
103501-474 103501-476
BclI-HF, New England Biolabs
Enzymes
A high fidelity restriction endonuclease that recognizes the sequence T^GATCA. High Fidelity (HF) Restriction Enzymes have 100% activity in CutSmart Buffer; single-buffer simplicity means more straightforward and streamlined sample processing.
  • Engineered for improved performance
  • 100% activity in CutSmart Buffer (over 215 enzymes are available in the same buffer)
  • Reduced star activity
HF enzymes are all Time-Saver qualified and can therefore cut substrate DNA in 5 to 15 with the flexibility to digest overnight without degradation to DNA. Engineered with performance in mind, HF restriction enzymes are fully active under a broader range of conditions, minimizing off-target products, while offering flexibility in experimental design.
Delivery information: Supplied with 1 vial of Gel loading dye, purple (6X).
Order Now
SPECIFICATIONS
Enzyme name BclI-HF
Environmentally Preferable
Source
Species
Concentration 20000 units/ml