Recombinant 1-2,4,6 Fucosidase O
Supplier: New England Biolabs (NEB)
P0749L P0749S
76196-416EA 911 CAD
76196-416 76196-418
Recombinant 1-2,4,6 Fucosidase O
Enzymes
a1-2,4,6 Fucosidase O is a broad specificity exoglycosidase that catalyzes the hydrolysis of terminal a1-2, a1-4 and a1-6 linked fucose residues from oligosaccharides. a1-2,4,6 Fucosidase O cleaves a1-6 fucose residues more efficiently than other linkages.
  • Recombinant enzyme with no detectable endoglycosidase or other exoglycosidase contaminating activity
  • Glycerol -free for optimal performance in HPLC and mass spectrometry analysis
  • ≥95% purity, as determined by SDS-PAGE and intact ESI-MS
  • Optimal activity and stability for up to 12 months