Thermolabile USER II Enzyme
Supplier: New England Biolabs (NEB)
M5508L
M5508S
76196-398EA
661
CAD
76196-398
76196-400
Thermolabile USER II Enzyme
Enzymes
Thermolabile USER (Uracil-Specific Excision Reagent) II Enzyme generates a single nucleotide gap at the location of a uracil residue. It can be 100% inactivated at temperatures >65 °C.
- Directional RNA SeqNEBNext adaptor cleavage
- USER cloning