TelN Protelomerase
Supplier: New England Biolabs (NEB)
M0651S
76196-388EA 124 CAD
76196-388
TelN Protelomerase
Enzymes
TelN Protelomerase, from phage N15, cuts dsDNA at a TelN recognition sequence (56 bp) and leaves covalently closed ends at the site of cleavage.
  • Isolated from phage N15
  • Cuts dsDNA at a TelN recognition sequence (56 bp)
  • Leaves covalently closed ends at the site of cleavage
  • Tel N is an Enzyme for Innovation (EFI). EFI is a project initiated by NEB to provide unique enzymes