NEBNext Single Cell/Low Input cDNA Synthesis and Amplification Module, New England Biolabs
Supplier: New England Biolabs (NEB)
E6421L E6421S
CA76293-218EA 3682 CAD
CA76293-218 CA76293-220
NEBNext Single Cell/Low Input cDNA Synthesis and Amplification Module, New England Biolabs
Nucleic Acid Reagents RT-PCR and cDNA Reagents
The NEBNext Single Cell/Low Input cDNA Synthesis and Amplification Module uses a template switching method to generate full length cDNAs directly from single cells or 2 pg to 200 ng RNA.
  • Experience unmatched detection of low abundance transcripts
  • Rely on consistent transcript detection for a wide range of input amounts and sample types
  • Observe uniform transcript coverage, regardless of input amount or sample type
  • Use with a variety of RNA inputs, including cultured or primary cells, or total RNA
This unique workflow enables generation of high quality sequencing data from a broad range of inputs, and superior transcript detection, while providing reliably consistent performance.
Order Now
SPECIFICATIONS
Environmentally Preferable
Application
Learn more