VWR® Vertical Racks with Spring Lock
Supplier: CRYSTAL TECHNOLOGY & INDUSTRIES
MCF-9-2-S MCF-10-2-S MCF-11-2-S MCF-12-2-S MCF-13-2-S MCF-14-2-S MCF-15-2-S
76051-582EA 98.86 CAD
76051-582 76051-584 76051-586 76051-588 76051-590 76051-592 76051-594
VWR® Vertical Racks with Spring Lock
Racks Cryogenic Box Racks
Vertical racks for mini 2" boxes with spring lock.
  • Stainless steel
  • Double welds
  • Smoothed edges
  • Lifetime warranty
Freezer Racks are made to the highest levels of quality with pharmaceutical grade stainless steel, double welds, smoothed edges and an unconditional lifetime warranty.