2-Propanol ≥99.5% Dehydrant, Epredia™ for histology

Supplier: RICHARD-ALLAN SCIENTIFIC CA
Danger

Synonyms: Isopropyl rubbing alcohol, Isopropanol, 2-Hydroxy propane, IPA, Dimethyl carbinol, Isopropyl alcohol, Propan-2-ol

9511 9515
CA72060-068CS 1212.7 CAD
CA72060-068 CA72060-070
2-Propanol ≥99.5% Dehydrant, Epredia™ for histology
2-Propanol
Formula: (CH₃)₂CHOH
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00011674
CAS Number: 67-63-0
UN: 1219
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR