SafeBasics® Bouffant Caps, Medicom
Supplier: Medicom
8001 8003 8004
CA76457-802CS 157.72 CAD
CA76457-802 CA76457-804 CA76457-806 CA76457-808
SafeBasics® Bouffant Caps, Medicom
Disposable Garments
Fluid-resistant hair and head protection.
  • Breathable, low-linting design
  • Latex-free elastic headband
  • Fluid repellent fabric for optimal protection
  • Available in sky blue and white
  • Single-use and disposable
Non sterile bouffant caps are intended to be worn by healthcare personnel during various procedures to protect patient/materials and the operating personnel from the transfer of microorganisms and particulates.
Packaging: Caps are packaged 100 per box, 5 boxes per case.
Order Now
SPECIFICATIONS
Fabric Spunbonded Polypropylene
Environmentally Preferable
Gender Unisex
Garment application Laboratory