SafeWear™ High Performance Lab Coats, Medicom
Supplier: Medicom
8107-A 8107-B 8107-C 8107-D 8107-E 8107-F 8109-A 8109-B 8109-C 8109-D 8110-A 8110-B 8110-C 8110-D 8108-D 8112-A 8112-B 8112-C 8112-D 8117-A 8117-B 8117-C 8117-D 8119-A 8119-B 8119-C 8119-D
CA76457-766CS 832.77 CAD
CA76457-766 CA76457-768 CA76457-770 CA76457-772 CA76457-774 CA76457-776 CA76457-778 CA76457-780 CA76457-782 CA76457-784 CA76457-786 CA76457-788 CA76457-790 CA76457-792 CA76544-746 CA76545-408 CA76545-410 CA76545-412 CA76545-414 CA76545-398 CA76545-400 CA76545-402 CA76545-404 CA76545-416 CA76545-418 CA76545-420 CA76545-422
SafeWear™ High Performance Lab Coats, Medicom
Disposable Garments
Length complies with OSAP recommendation for knee-length coverage.
  • Available in a wide range of styles, colors, and sizes
  • Comply with recommended OSHA best practices
  • Made from high-quality 45 g/m² SMS fabric
  • Detailed finishing, 3 pocket design, with front snap closure
  • Disposable, single-use garment
  • Latex-free
  • Thoughtful designs offer ease of movement, as well as secure closure and easy removal
  • Premium materials provide superior protection, reliability, and durability
  • Lightweight, soft, supple, fabrics keep wearers cool and comfortable
  • SafeWear™ protective apparel complies with recommended OSHA best practices
SafeWear™ premium protective apparel is made from top quality materials and designed to provide reliable barrier protection without sacrificing comfort or style. The High Performance Lab Coat is a procedure SMS garment designed to protect health professionals and patients from cross contamination. Made of non-woven material with front closure and snap buttons.These coats feature a knitted collar and cuffs, as well as two slits in the back.
Certifications: Fluid-resistant fabric meets AAMI PB70 Level 2 standards.
Packaging: Lab coats are packaged 12/bag, 5 bags/case.