SafeWear™ Hipster Jacket™, Medicom
Supplier: Medicom
8101-A 8101-B 8101-C 8101-D 8101-E 8101-F 8103-A 8103-B 8103-C 8103-D 8105-A 8105-B 8105-C 8105-D 8115-A 8115-B 8115-C 8115-D 8116-A 8116-B 8116-C 8116-D 8118-A 8118-B 8118-C 8118-D
CA76457-722CS 832.77 CAD
CA76457-722 CA76457-724 CA76457-726 CA76457-728 CA76457-730 CA76457-732 CA76457-734 CA76457-736 CA76457-738 CA76457-740 CA76457-750 CA76457-752 CA76457-754 CA76457-756 CA76544-830 CA76544-832 CA76544-834 CA76544-836 CA76544-838 CA76544-840 CA76544-842 CA76544-844 CA76544-846 CA76544-848 CA76544-850 CA76544-852
SafeWear™ Hipster Jacket™, Medicom
Disposable Garments
Reliable protection without sacrificing comfort or style.
  • Available in a wide range of styles, colors, and sizes
  • Comply with recommended OSHA best practices
  • Made from high-quality 45 g/m² SMS fabric
  • Detailed finishing, 3 pocket design, with front snap closure
  • Disposable, single-use garment
  • Latex-free
  • Thoughtful designs offer ease of movement, as well as secure closure and easy removal
  • Premium materials provide superior protection, reliability, and durability
  • Lightweight, soft, supple, fabrics keep wearers cool and comfortable
  • SafeWear™ protective apparel complies with recommended OSHA best practices
SafeWear™ premium protective apparel is made from top quality materials and designed to provide reliable barrier protection without sacrificing comfort or style. The Hipster Jacket™ is a procedure SMS garment designed to protect health professionals and patients from cross contamination. Made of non-woven material with front closure and snap buttons.These jackets feature a knitted collar and cuffs.
Certifications: Fluid-resistant fabric meets AAMI PB70 Level 2 standards.
Packaging: Jackets are packaged 12 per bag, 5 bags per case.