Cloned Pfu DNA Polymerase Alternative Detergents, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES (GENOMICS) CA
600355 600357
CAST600353EA 215.43 CAD
CAST600353 CAST600355 CAST600357
Cloned Pfu DNA Polymerase Alternative Detergents, Agilent Technologies
Nucleic Acid Reagents End-point PCR Enzymes and Kits
Cloned Pfu DNA Polymerase for high fidelity and difficult, GC-Rich PCR.
  • Highly thermostable Pfu DNA polymerase possessing 3' to 5' exonuclease proofreading activity that enables the polymerase to correct nucleotide-misincorporation errors (1 error per 770000 bp)
  • Reformulated buffer system for increased economy with the same high performance as the original Pfu DNA polymerase
  • Blunt ends
Using Cloned Pfu DNA polymerase in PCR experiments results in blunt-ended products, which are ideal for cloning into blunt-ended vectors. Cloned Pfu DNA Polymerase, derived from the hyperthermophilic archaea Pyrococcus furiosus exhibits superior thermostability and proofreading properties.
Caution: For research use only. Not for use in diagnostic procedures.
Order Now
SPECIFICATIONS
Environmentally Preferable
Application
Learn more