PCR and qPCR Consumables, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES (GENOMICS) CA
410186 600567 410088 410092 410152 401333 401334 410094
CA76204-934PK 192.58 CAD
CA76204-934 CA76204-936 CA76204-938 CA97066-460 CA99900-140 CA99900-152 CA99901-852 CA99901-854 CA99901-856 CA99901-860 CA99901-868 CAST401333 CAST401334 CAST410094
PCR and qPCR Consumables, Agilent Technologies
Thermal Cyclers
High-quality well plate supplies and accessories will enhance your PCR and qPCR workflow and analysis.
Agilent AriaMx low profile strip tubes are optimized for PCR and qPCR applications. The tubes are validated for optimal performance with Mx3005P, Mx3000P, and the AriaMx real time PCR Systems. Part of the qPCR Plastics and Accessories, the products include the perfect fit frame, tubes, and caps.
Additional products available include the 96-well semiskirted polypropylene PCR plate compatible with the optical Strip Caps, the StrataCoolers LP Benchtop Cooler, and StrataCooler cryoMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable