PCR Plates, Globe Scientific
Supplier: Globe Scientific
PCR-384-ABI PCR-384-ABIW PCR-384-ROC PCR-384-ROCW PCR-FS-02 PCR-HS-01W PCR-HS-02 PCR-HS-FL02 PCR-HS-RR01 PCR-HS-RR02 PCR-NS-01 PCR-NS-02
76468-990BX 87.17 CAD
76468-990 76468-992 76468-994 76468-996 76468-998 76469-000 76469-002 76469-004 76469-006 76469-008 76469-010 76469-012
PCR Plates, Globe Scientific
Microplates PCR Plates
Globe Scientific® PCR plates are offered in a variety of formats including no skirt, half skirt and full skirt variants in 0.1 ml (low profile) and 0.2 ml volumes. Available 384 well variants round out this expansive offering.
  • Ultra-thin wall design (only 0.2 mm thick) for optimal temperture transfer
  • Ultra-clear wells for easy sample visualization
  • Easy-to-read alphanumeric indexing
  • Manufactured from high-clarity, ultra-pure polypropylene (meets FDA 21 CFR 177.1520 & USP Class VI)
  • Certified free of detectable DNase, RNase, Human DNA, PCR Inhibitors and
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable