PCR Tube and Cap Strips, Globe Scientific
Supplier: Globe Scientific
PCR-02DC PCR-02FC PCR-02S PCR-DL-01F PCR-DL-01FW PCR-DL-02F PCR-QS-02D PCR-QS-02F PCR-STR-01F PCR-STR-01FW PCR-STR-02D PCR-STR-02F
76468-966BX 55.66 CAD
76468-966 76468-968 76468-970 76468-972 76468-974 76468-976 76468-978 76468-980 76468-982 76468-984 76468-986 76468-988
PCR Tube and Cap Strips, Globe Scientific
Tubes PCR Tubes
The Diamond® PCR Strip tubes and caps offering includes DiamondLink™ strips which feature extremely strong links between wells for ease of handling and QuickSnap™ strips that easily snap apart for customization into smaller strips or use in multiple analyses.
  • Ultra-thin wall design (only 0.2 mm thick) for optimal temperature transfer
  • Certified free of detectable DNase, RNase, Human DNA, PCR inhibitors and tested Pyrogen-free
  • DiamondLink™ strips include 45° nested attached individual caps and strong links between tubes for easy handling
  • QuickSnap™ also feature 45° nested attached individual caps but have breakable links between tubes for easy customization
Both DiamondLink™ and Quicksnap™ caps are attached at 45º angles to prevent interference with one another when used in SBS format racks. Separate tube and cap strips are available individually or in combination packs. Low profile and white options are also available for smaller reactions and optimal qPCR performance.