EZ Air Tyvek® and Saran™-Coated Tyvek® PAPR Hood Assemblies, Allegro®
Supplier: Allegro
9933 9933-S
76437-508EA 2085.69 CAD
76437-508 76437-510
EZ Air Tyvek® and Saran™-Coated Tyvek® PAPR Hood Assemblies, Allegro®
Respirators Respiratory Systems
The EZ Air PAPR Hooded Respirators are complete particulate systems.
  • Fully disposable hood made of poly-coated DuPont Tyvek® or Saran™-coated Tyvek® with bonded/taped seams
  • Disposable one-piece suspension
  • NIOSH approved HEPA filter (42 CFR, Part 84)
  • Blower is rugged, lightweight ABS (blower, filter, and battery weight combined: 2.62 lbs.)
  • 7.4V rechargeable Lithium-ion battery
  • 110 to 240 VAC, 50 to 60 Hz smart charger
  • Waist belt features a fire retardant outer material
The EZ Air Powered Air Purifying Respirator (PAPR) is a complete particulate system intended for both industrial and healthcare workers where combined respiratory, eye, and face protection are required.
It is a motorized, Lithium-Ion battery operated system that provides a constant supply of fresh, filtered air over the user’s face. At an APF level of 25, it creates an optimal protective pressure inside the respirator hood, keeping out harmful particulates. It’s also extremely lightweight, weighing under 4 pounds in total.
Recommended applications are facility maintenance and healthcare facilities.
Certifications: NIOSH approved.
Ordering information: Complete PAPR assembly includes choice of either poly-coated Tyvek® or Saran™-coated Tyvek® hood, breathing tube, blower, battery, waist belt, charger, flow tester, and HEPA filter.