Ward's Drying Rack Set
Supplier: Heathrow Scientific
120828 120792 120793 120813 120862W
470331-206CS 692.85 CAD
470331-206 470331-208 470331-210 470331-212 470331-770
Ward's Drying Rack Set
Racks Draining Racks
The most versatile drying rack on the market today!
  • Features channels designed for more configurations and to protect against breakage
  • Drying Basket to keep small items and slotted to hold pipettes safe while drying
  • SureGrip Hook with Patent-pending design to securely grips glassware into place
The Ward's drying rack can be used on counters with integrated hinged stand, or walls with included guide and hardware. Made of durable polystyrene, the rack can hold up to 77 pounds.
Delivery information: Includes 72 pegs (36 small, and 36 large), and twoMore Product Information