AccuScript High-Fidelity Reverse Transcriptase, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES (GENOMICS) CA
200820 600089
CA97066-438KT 826.07 CAD
CA97066-438 CA99902-640 CA99902-688 CAST200820 CAST600089
AccuScript High-Fidelity Reverse Transcriptase, Agilent Technologies
Nucleic Acid Reagents RT-PCR and cDNA Reagents
AccuScript High-Fidelity Reverse Transcriptase delivers the highest RT accuracy currently available.
  • MMLV-RT derivative combined with 3 to 5’ exonuclease reduces errors
  • High yields of full length cDNA up to 20 kb;RT-PCR kit synthesises cDNA from 10 to 1000 ng of total RNA or 0.1 to 10 ng of poly(A)+ RNA
  • Stringent quality control procedures ensure high yields of premium cDNA
AccuScript High-Fidelity Reverse Transcriptase (RT) delivers 3.7 times more accuracy than other commercially available RTs and 6.6 times more accuracy than AMV RT. The AccuScript PfuUltra II RT-PCR kit provides a complete system in applications that demand high fidelity, specificity, and throughput.
Caution: For research use only. Not for use in diagnostic procedures.