BIC® Gamma Irradiated Cleanroom Pens, Keystone Cleanroom Products
Supplier: KEYSTONE ADJUSTABLE CAP CO., INC.
S01PENBLK S01PENBLUE S02PENBLK S02PENBLUE S05PENBLK S05PENBLUE S10PENBLK S10PENBLUE
76451-904CS 3566.03 CAD
76451-904 76451-906 76451-916 76451-918 76451-928 76451-930 76451-940 76451-942
BIC® Gamma Irradiated Cleanroom Pens, Keystone Cleanroom Products
Pens Cleanroom Pens
Ready to Use gamma irradiated pens and markers eliminate the risks associated with surface sanitization procedures.
  • Gamma irradiated
  • Lot controlled
  • Ready to use
  • Packaged in easy to open Tyvek® peel pouches
  • Minimize contamination risks
  • Simplifies conformance to SOP's
  • Eliminate the need for wiping down contaminated writing intruments
Keystone CRP offers gamma irradiated pens and markers for the convenience of pharmaceutical cleanroom operations. These are a 'Ready to Use Option' when sterilization on-site is not possible; or surface sterilization isMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable