FlexTech™ Y9219 Ultra Lightweight Liners, Wells Lamont
Supplier: Wells Lamont
Y9219S Y9219M Y9219L Y9219XL Y9219XXL
76470-992PK 359.5 CAD
76470-992 76470-994 76470-996 76470-998 76471-000
FlexTech™ Y9219 Ultra Lightweight Liners, Wells Lamont
Gloves
Superior cut protection with ultimate dexterity, tactility, and comfort.
  • PU coated
  • High abrasion resistance
  • Touchscreen compatible
  • High puncture performance
The Y9219 has an ultra lightweight liner that provides excellent dexterity, with an ANSI cut level A9 for superior cut protection to reduce hand injuries. These gloves offer increased abrasion resistance and durability for prolonged use.
Certifications: ANSI cut A9, abrasion level 4, puncture level 4.
Ordering information: Sold by the dozen pairs.
Order Now
SPECIFICATIONS
Ambidextrous/Hand-specific Ambidextrous
Environmentally Preferable
Disposable / reusable Reusable
Latex/Latex-free Latex-free
Powdered/Powder-free Powder-free
Gauge 18