NEBNext® Multiplex Oligos for Illumina (Unique Dual Index UMI Adaptors DNA Set 1), New England Biolabs
Supplier: New England Biolabs (NEB)
E7395S E7395L
CA76470-778EA 1076 CAD
CA76470-778 CA76470-780
NEBNext® Multiplex Oligos for Illumina (Unique Dual Index UMI Adaptors DNA Set 1), New England Biolabs
Nucleic Acid Reagents Next Generation Sequencing Reagents
NEBNext® Multiplex Oligos for Illumina are an essential piece of the NEBNext® suite of library preparation products, available in several different configurations for use with NEBNext® products or other standard Illumina-compatible library preparation protocols.
  • Index adaptors enable PCR-free library preparation
  • More-sensitive detection of low-frequency single-nucleotide variants (SNVs)
  • Supports multiplexing for higher throughputs
This set was designed and optimized for use in DNA sequencing workflows, including PCR-free. These unique dual index adaptors also include a unique molecular identifier (UMI) sequence to enable the identification and removal of PCR errors or duplicates from amplified libraries. Supplied as 96 unique, pre-annealed index adaptors with UMIs, packaged in a single-use 96-well plate with a pierceable foil seal. Universal primers are included for applications requiring PCR amplification.