Clamp-On Breath Shields, Eagle MHC
Supplier: Eagle Group
BGOC-2424 BGOC-2436 BGOC-2448 BGOC-2460 BGOC-3030 BGOC-3036 BGOC-3048 BGOC-3060
76452-116EA 311.62 CAD
76452-116 76452-118 76452-120 76452-122 76452-124 76452-126 76452-128 76452-130
Clamp-On Breath Shields, Eagle MHC
Shields Safety Shields
Add a layer of protection to help maintain social distancing and provide a physical barrier to protect staff and customers.
  • Shields clamp onto table edge surface
  • ⁷/₃₂" thick durable plexiglass
  • Clamp range is 1 to 1.5" (can be customized)
  • Stainless clamps have protective rubber interior to protect table surface
Shields clamp onto table edge surface. Individual shields can be configured based on layout requirements. Clamps accommodate surfaces from 1 to 1.5" thick.