Distillation Condenser, Ace Glass
Supplier: Ace Glass
6603-02 6603-04 6603-06
76470-472EA 1565.1 CAD
76470-472 76470-474 76470-476
Distillation Condenser, Ace Glass
Condensers
Liebig condenser with vacuum monitoring port that, when mated to our 74670 family of columns completes a short path distillation head.
  • Two-piece design simplies cleaning
  • Ball and socket joint allow for adjustability
  • High-quality borosilicate glass construction
  • Made in the USA
Two-piece design simplifies cleaning and our 35/25 O-ring ball and socket joint connection allows for some adjustability over a standard taper joint configuration.
Ordering information: For a complete short path distillation head order with 76470-468 or 76470-470 distillation column.