Fume Hood Worktops, Kewaunee
Supplier: Kewaunee
VBTS012448 VBTS012460 VBTS012472 VBTS012496 VBTS013048 VBTS013060 VBTS013072 VBTS013096 VBTS013648 VBTS013660 VBTS013672 VBTS013696
KSVBTS012448EA 15920.66 CAD
KSVBTS012448 KSVBTS012460 KSVBTS012472 KSVBTS012496 KSVBTS013048 KSVBTS013060 KSVBTS013072 KSVBTS013096 KSVBTS013648 KSVBTS013660 KSVBTS013672 KSVBTS013696
Fume Hood Worktops, Kewaunee
Fume Hoods and Enclosures Chemical Fume Hoods
Fume hood worktop, 304L stainless steel without cupsink.
Venturi bench hoods require a work top that must be ordered separately. Work tops are available in either kemresin or stainless steel, are dished ¹/₂" to retain spillage, and incorporate a 2" wide safety rim at the front. They may include cutouts for cupsinks or vent pipes when specified with the cutout option. Cupsinks must be ordered separately. Venturi floor founted hoods may be ordered with a removable work shelf and/or a work floor that must be ordered separately. Both are available in either kemresin or stainless steel, areMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable