Spherical Reaction Flasks with Short Neck, Ace Glass
Supplier: Ace Glass
6533-08 6533-13
76470-464EA 812.3 CAD
76470-464 76470-466
Spherical Reaction Flasks with Short Neck, Ace Glass
Flasks Round Bottom Flasks
High-quality borosilicate glass spherical reaction flasks.
  • Duran O-ring flange
  • 150 mm flange
  • Short neck
Reaction flasks featuring a ground O-ring Duran flange with a short neck.