Glass Head with Duran® Flange and Three Necks, Ace Glass
Supplier: VWR International
76470-460 76470-462
76470-460EA 927.23 CAD
76470-460 76470-462
Glass Head with Duran® Flange and Three Necks, Ace Glass
Lids Reaction Vessel Lids
These Duran® flange glass heads are made with high-quality borosilicate glass.
  • Duran® O-ring flange
  • 24/40 center neck
  • (2) 24/40 side necks
Featuring three in-line standard taper joints with a Duran® O-ring flange to ensure a tight seal with any of the matching 150 mm flanged vessels.
Order Now
SPECIFICATIONS
Environmentally Preferable