VacTrap-G® (Glass) Vacuum Trap System, Foxx Life Sciences®
Supplier: Foxx Life Sciences
30R-1101-FLS 30R-2001-FLS 30R-2101-FLS 30R-2201-FLS 30R-3101-FLS 30R-5001-FLS 30R-5201-FLS 30R-A201-FLS 30R-C501-FLS
76449-392EA 693.91 CAD
76449-392 76449-394 76449-396 76449-398 76449-400 76449-402 76449-404 76449-406 76449-408
VacTrap-G® (Glass) Vacuum Trap System, Foxx Life Sciences®
Vacuum Traps
Vactrap™ G (Glass) vacuum trap system, adher to the strict CDC recommendation for laboratory safety.
  • Vactrap™ G includes the patented VersaCap® closure for tangle-free opening and closing
  • Polypropylene VersaCap® has the highest temperature rating in the industry, 142 °C
  • A 0.2 μm PTFE inline vent filter retains any aerosolized bacteria and protects the house vacuum or pump
  • The entire assembly is nested neatly in a secondary container with a small footprint and incredible stability against tipping
  • Pinch clamp allows vacuum adjustment
  • Foxx EZFlow® TPE
More Product Information