Nalgene™ Vacuum Tubing, Ace Glass
Supplier: Ace Glass
12691-03 12691-05 12691-07
76470-478CS 614.78 CAD
76470-478 76470-480 76470-482
Nalgene™ Vacuum Tubing, Ace Glass
Tubing Transfer Tubing
Nalgene™ Non-Phthalate Clear Plastic PVC Vacuum Tubing performs like classic Nalgene™ 180 tubing but is free from orthophthalate DEHP plasticizers.
  • Non-Phthalate PVC
  • Lab-grade, autoclavable
  • Resists hardening and discoloration
  • Marked with int.Ø and regulatory compliance
Withstands a full vacuum (29.9 in Hg) at room temperature. Equivalent to Tygon™ E-3603 tubing.
Certifications: Complies with FDA CFR21 for food packaging, USP VI, Cytotoxicity, REACH, Latex-free, RoHS, and complies with California proposition 65.