VWR® Vertical Racks
Supplier: VWR International
76051-508 76051-574 76051-576 76051-578 76051-580
76051-508EA 34.08 CAD
76051-508 76051-574 76051-576 76051-578 76051-580
VWR® Vertical Racks
Racks Cryogenic Box Racks
Vertical freezer racks for 2" boxes.
  • Stainless steel
  • Double welds
  • Smoothed edges
  • Lifetime warranty
Freezer Racks are made to the highest levels of quality with pharmaceutical grade stainless steel, double welds, smoothed edges and an unconditional lifetime warranty.