VWR® Upright Freezer Drawer Racks
Supplier: VWR International
76051-430 76051-432 76051-434 76051-436 76051-438
76051-430EA 424.07 CAD
76051-430 76051-432 76051-434 76051-436 76051-438
VWR® Upright Freezer Drawer Racks
Racks Cryogenic Box Racks
Upright freezer drawer rack for 2" boxes and 15 or 50 ml centrifuge tubes.
  • Stainless steel
  • Double welds
  • Smoothed edges
  • Lifetime warranty
These freezer racks are made to the highest levels of quality with pharmaceutical grade stainless steel, double welds, smoothed edges and an unconditional lifetime warranty.