Pliers, Stress Relief, Excelta
Supplier: Excelta
554AD-US 554B-US 554EF-US 554TXB-US 554TX-US 900-88B 907-88 909-4 909-4B 554F-US 954A 554TB-US 554TXC-US 900-10 900-247 907-88C 954B
63042-588EA 781.56 CAD
63042-588 63042-596 63042-604 63042-610 63042-612 63043-010 63043-040 63043-046 63043-048 89411-956 89411-958 75880-626 75880-628 75880-652 75880-656 75880-660 75880-668
Pliers, Stress Relief, Excelta
Pliers
Designed to form stress relief for transistor and resistor leads.
  • Stress relief on transistors, resistors
  • Ergonomic grips for comfort
  • Double leaf springs for ease of use
All of these forming pliers provide stress relief for transistor and resistor leads. They feature an ergonomically molded antimicrobial grip (10¹⁰ ohms/sq surface resistivity) with dual leaf springs. The tools are made of carbon steel.