Pliers, Lock-In Lead Forming, Excelta
Supplier: Excelta
900-3RB 554-3RT-US 554-3RB-US 900-3RT
89411-964EA 1112.83 CAD
89411-964 89411-966 63042-584 63042-586
Pliers, Lock-In Lead Forming, Excelta
Pliers
This series form an 'S' bend on a component lead, enabling the component to stay in place when soldering.
  • Will form lead and lock it into place on PC board
  • Ergonomic grips for comfort
  • Double leaf springs for ease of use
These lock-in lead forming pliers will form an 'S' bend on a component lead, enabling the component to stay in place when soldering. They are recommended for 0.025" diameter or smaller leads and wires. It features a ergonomically molded antimicrobial grip (10¹⁰ ohms/sq surface resistivity) with dual leaf springs. These tools are made of carbon tool steel with flame hardened jaws.