Pliers, Wide Nose, Excelta
Supplier: Excelta
508B-US 508D-US 508-US 508C-US
63042-492EA 795.01 CAD
63042-492 63042-494 63042-496 75880-924
Pliers, Wide Nose, Excelta
Pliers
The 508 series will straightens multiple lead wires in one easy operation.
  • Wide nose ideal for large items
  • Ergonomic grips for comfort
  • Double leaf springs for ease of use
These wide nose straightening pliers uniquely straighten multiple leads in one easy operation. The jaw inserts are factory replaceable and available in carbon steel, brass, or acetal. The overall length is 5.5". It features a ergonomically molded antimicrobial grip (10¹⁰ ohms/sq surface resistivity) with dual leaf springs. The shank is made of carbon steel.