Pliers, Multiple Lead Forming, Excelta
Supplier: Excelta
500-210-US 554AJ-US 554N-US 554-US 907-IC 954 500-12-US 500-15-US 500-210A-US 554BJ-US 500-210C-US
63042-414EA 787.96 CAD
63042-414 63042-590 63042-608 63042-614 63043-044 63043-068 75880-608 75880-610 75880-616 75880-622 75880-922
Pliers, Multiple Lead Forming, Excelta
Pliers
These are ideal to bend the leads on LEDs and other multiple lead components consistantly.
  • Multi lead forming
  • Ergonomic grips for comfort
  • Double leaf springs for ease of use
The multiple lead forming plier is designed to bend the leads on LEDs and other multiple lead components. It features an ergonomically molded antimicrobial grip (10¹⁰ ohms/sq surface resistivity) with dual leaf springs. The tool is made of carbon steel.