Pliers, Transistor Former, Excelta
Supplier: Excelta
500-103-US 500-104B-US 500-104-US 907-104 907-4 500-11-US 500-103B-US 907-104A 907-210A
63042-390EA 1161.54 CAD
63042-390 63042-392 63042-394 63043-022 63043-030 89411-960 75880-614 75880-662 75880-664
Pliers, Transistor Former, Excelta
Pliers
These tools are ideal for forming leads on a TO-220 component in one easy operation.
  • Specifically designed for lead forming transistors
  • Ergonomic grips for comfort
  • Double leaf springs for ease of use
These transistor forming plier are designed to form the leads on a variety of transistors. They feature an ergonomically molded antimicrobial grip (10¹⁰ ohms/sq surface resistivity) with dual leaf springs. They are made of carbon tool steel.