Pliers, Miniature Self-Opening, Excelta
Supplier: Excelta
381 382 383 384 385 386
63042-076EA 90.58 CAD
63042-076 63042-078 63042-080 63042-082 63042-084 63042-086
Pliers, Miniature Self-Opening, Excelta
Pliers
Excellent when working in very difficult and confined areas.
  • Miniature for use under a scope
  • Cleanroom safe, stainless steel
  • Self-opening, spring loaded for ease of use
These self opening miniature pliers are excellent when working in very difficult and confined areas. The overall length is 3" and the jaws are ¹/₄". They are made of stainless steel. They are spring loaded and normally open.