Pliers, Needle Nose, Excelta
Supplier: Excelta
2647 2647D 2847 2847D 2847L 2847LD 2910D 2905 2647SD 2847-CR 2847D-CR 2847SL 2905D
63041-724EA 66.79 CAD
63041-724 63041-726 63041-810 63041-812 63041-814 94024-206 75880-682 76245-086 76246-954 76471-586 76471-610 76471-612 76471-614 76471-618
Pliers, Needle Nose, Excelta
Pliers
The tapered jaws on these pliers allow better accessibility in tight places.
  • Needle nose for tight areas
  • Ergonomic handle, dual leaf springs
  • Stainless steel construction-corrosion resistant
  • Different sizes depending on application
Needle nose pliers have tapered noses for better accessibility in tight places. The noses aren't quite as strong as the 'Chain Nose' pliers due to the taper. Each of these is made of AISI 440 stainless steel to prevent oxidation. All include two leaf springs for more ergonomic control. They are offered in a small, medium or large head side, depending on the application. The 'D' specifies serrations on the tips for better control.