Pliers, Flat Nose, Excelta
Supplier: Excelta
2642 2842 2842D 2842L 2818 2818-CR 2842-CR 2842D-CR
63041-716EA 62.88 CAD
63041-716 63041-794 63041-796 63041-798 94024-202 76471-588 76471-594 76471-596
Pliers, Flat Nose, Excelta
Pliers
Flat nose pliers are designed for straightening wires or component leads.
  • Flat nose for straightening wire, flashing
  • Ergonomic handle, dual leaf springs
  • Stainless steel construction-corrosion resistant
  • Different sizes depending on application
Flat nose pliers are made of AISI 440 stainless steel and include two leaf springs for more ergonomic control. They are offered in a small, medium or large head, depending on the application. They are most commonly used to straighten wires or flanges.