HALO® PFAS Solutions, ENVIROCLASS, HPLC Columns, Advanced Materials Technology
Supplier: Advanced Materials Technology
92812-413 92812-613 92812-713 92812-913 92813-413 92813-613 92813-713 92813-913 92113-415 92114-415
76469-452EA 1562.37 CAD
76469-452 76469-454 76469-456 76469-458 76469-460 76469-462 76469-464 76469-466 76469-468 76469-470
HALO® PFAS Solutions, ENVIROCLASS, HPLC Columns, Advanced Materials Technology
Chromatography Columns
Designed for separation of novel and legacy short chain and long chain PFAS compounds containing branched and linear isomers, and with EPA methodology requirements in mind, HALO® PFAS offers a holistic solution.
  • Application assured through method qualified lot analysis
  • Optimal 2.7 µm Fused-Core® particle for rugged, reliable performance delivering high efficiency, low back pressure separations
  • Endcapped alkyl phases for high sensitivity (no bleed) LCMS analysis
With both a PFAS-specific delay column optimized to prevent background PFAS contaminationMore Product Information